Learn More
Hampton Research ClearSeal Film 100 pack
SDP
Catalog No. 500214
Encompass
ClearSeal Film fits all SBS format 96 well plates. Minimal evaporation, minimal cross-well contamination, easy to handle, and optically clear. Single coated, 2 mil (0.05 mm), optically clear polyolefin film with pressure-sensitive silicone-based adhesive. Supplied as a 141 mm x 78 mm film with white polyester release liner (backing). Actual seal area after end tabs removed is 120 x 78 mm Seals well to polypropylene, polystyrene, and polycarbonate plates. Compatible with aqueous and organic solvents. Suitable for use between -70 and 110°C. For best seal, the plate must be sealed with the Film Sealing Paddle HR4-513. ClearSeal Film has a limited shelf life. Must be used before expiration date printed on box.
Catalog No. 50-021-4 Supplier Hampton Research Supplier No. HR4521
ClearSeal Film fits all SBS format 96 well plates. Minimal evaporation, minimal cross-well contamination, easy to handle, and optically clear. Single coated, 2 mil (0.05 mm), optically clear polyolefin film with pressure-sensitive silicone-based adhesive. Supplied as a 141 mm x 78 mm film with white polyester release liner (backing). Actual seal area after end tabs removed is 120 x 78 mm Seals well to polypropylene, polystyrene, and polycarbonate plates. Compatible with aqueous and organic solvents. Suitable for use between -70 and 110°C. For best seal, the plate must be sealed with the Film Sealing Paddle HR4-513. ClearSeal Film has a limited shelf life. Must be used before expiration date printed on box.