Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec ClearSteel, Stainless Steel Cleaning and Polishing Wipes
SDP

Catalog No. 22444301 Shop All Mopec Products
Change view
Click to view available options
Product Type:
Wipes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
22-444-301 Wipes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-444-301 Supplier Mopec Supplier No. BE039
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mopec ClearSteel, Stainless Steel Cleaning and Polishing Wipes allow users to clean soiled area while bringing out the natural luster in the equipment and leaving a protective covering that repels water, fingerprints and dust.

  • Specifically designed to be used on all equipment such as workstations, cabinets, coolers and storage areas
  • Wipe in the direction of the stainless steel grain for best results

Specifications

Product Type Wipes
Color-coded No
Form Sachet Wipe
pH 8 to 9
Scent Unscented
Sterility Sterile
Appearance White
Container Type Canister
Content And Storage Store in a cool dry place
Description Cleaning Wipes
For Use With (Application) Cleaning, Polishing
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.