Learn More
ClearView Aluminum-Hematein Dye Lake Complex, StatLab
SDP
Catalog No. 22024792
Change view
Click to view available options
Quantity:
500 mL
Catalog No. Quantity
22-024-792 500 mL
1 options
Catalog No. 22-024-792 Supplier SLMP Llc Dba Statlab Medical Supplier No. MA0101211
Add to Cart
Add to Cart
It is a Hematoxylin Stain
  • A selective nuclear stain
  • Acts as a basic (positively charged) dye that binds to acidic (negatively charged) basophilic components, specifically the DNA and RNA within cell nuclei
  • Stains nuclear chromatin a dark blue or purple color
Specifications
Quantity 500 mL