Learn More
StatLab™ ClearView Eosin
SDP
Catalog No. STMA0101015
Change view
Click to view available options
Quantity:
500 mL
Catalog No. Quantity
STMA0101015 500 mL
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STMA0101015 Supplier StatLab™ Supplier No. MA0101015
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
An ideal counterstain for ClearView hematoxylin.
  • Produces enhanced staining of nuclear and non-nuclear cellular components and improved differentiation by graded alcohols in the staining sequence
  • Use with ClearView Hematoxylin
Specifications
Quantity 500 mL
Product Type Eosin Stain