Learn More
StatLab™ ClearView Hematoxylin
SDP
Catalog No. STMA0101010
Change view
Click to view available options
Quantity:
500 mL
Catalog No. Quantity
STMA0101010 500 mL
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STMA0101010 Supplier StatLab™ Supplier No. MA0101010
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
An ideal hematoxylin stain.
  • Stain produces enhanced staining of nuclear and non-nuclear cellular components and improved differentiation by graded alcohols in the staining sequence
  • Use with ClearView Eosin
Specifications
Quantity 500 mL
Product Type Hematoxylin Stain