Learn More
Marketlab ClearWave Ultrasound Gel Bottle
Description
- Premium, high-quality water-soluble gel
- Proprietary production method ensures gel is virtually free of micro-bubbles to reduce image artifact and improve transducer coupling
- Consistent viscosity proven in clinical trials
- Low-density, easy-to-squeeze poly bottles aid in dispensing and reduce waste
- Reduce risk of cross-contamination with built-in check valve opening and flip-top cap
- Five-year shelf life
Specifications
Specifications
| For Use With (Application) | Medium Viscosity Gel |
| Quantity | 12/Pk. |