Learn More
Cleatech SDPVC 32" FLOW HOOD W/ STND
SDP
Catalog No. 501527915
Encompass_Preferred
Portable Bench top Vertical Laminar Flow station, Work space 31.5"Wx 21.75"Dx 23.75"H , Clear Static Dissipative PVC construction, Clear Satic Dissipative PVC front tilt-up sash,110VAC, 50/60Hz. Adjustable speed Impeller, Backward Curve, 730 CFM @ 0.000-In,Including 30" Stand ,Including HEPA Filter , Including Built-in Fluorescent Light
Catalog No. 50-152-7915 Supplier Cleatech Supplier No. 10007DFLS
Add to Cart
Add to Cart
Request a Quote
Portable Bench top Vertical Laminar Flow station, Work space 31.5"Wx 21.75"Dx 23.75"H , Clear Static Dissipative PVC construction, Clear Satic Dissipative PVC front tilt-up sash,110VAC, 50/60Hz. Adjustable speed Impeller, Backward Curve, 730 CFM @ 0.000-In,Including 30" Stand ,Including HEPA Filter , Including Built-in Fluorescent Light