Learn More
Cleatech SDPVC 24" DUCTLESS FUME HOOD
SDP
Catalog No. 501527916
Encompass_Preferred
Portable 24" Width Bench top ductless exhaust hood, Work space 23"Wx 17.5"Dx 17"H , Polypropylene worksurface, Static-dissipative PVC walls, Clear Static-dissipative PVC front tilt-up sash,Polypropylene Fan/ Filter Housing. 110VAC, 60Hz. 155W Adjustable speed Impeller, Backward Curve, 1260 CFM @ 0.000-In,Including HEPA filter , Including Carbon filter
Catalog No. 50-152-7916 Supplier Cleatech Supplier No. 11002CHC
Portable 24" Width Bench top ductless exhaust hood, Work space 23"Wx 17.5"Dx 17"H , Polypropylene worksurface, Static-dissipative PVC walls, Clear Static-dissipative PVC front tilt-up sash,Polypropylene Fan/ Filter Housing. 110VAC, 60Hz. 155W Adjustable speed Impeller, Backward Curve, 1260 CFM @ 0.000-In,Including HEPA filter , Including Carbon filter