Learn More
Cleatech SDPVC 24" D/L HOOD W/ STND
SDP
Catalog No. 501527917
Encompass_Preferred
Portable 24" Width Bench top ductless exhaust hood, Work space 23"Wx 17.5"Dx 17"H , Polypropylene worksurface, Static-dissipative PVC walls, Clear Static-dissipative PVC front tilt-up sash,Polypropylene Fan/ Filter Housing. 110VAC, 60Hz. 155W Adjustable speed Impeller, Backward Curve, 1260 CFM @ 0.000-In, Including 30" Stand , Including HEPA filter , Including Carbon filter
Catalog No. 50-152-7917 Supplier Cleatech Supplier No. 11002CHCS
Portable 24" Width Bench top ductless exhaust hood, Work space 23"Wx 17.5"Dx 17"H , Polypropylene worksurface, Static-dissipative PVC walls, Clear Static-dissipative PVC front tilt-up sash,Polypropylene Fan/ Filter Housing. 110VAC, 60Hz. 155W Adjustable speed Impeller, Backward Curve, 1260 CFM @ 0.000-In, Including 30" Stand , Including HEPA filter , Including Carbon filter