Learn More
Cleatech SDPVC 32" DUCTLESS FUME HOOD
SDP
Catalog No. 501527918
Encompass_Preferred
Portable 32" Width Bench top ductless exhaust hood, Work space 31"Wx 17.75"Dx 17"H , Polypropylene worksurface, Static-dissipative PVC walls, Clear Static-dissipative PVC front tilt-up sash,Polypropylene Fan/ Filter Housing. 110VAC, 60Hz. 155W Adjustable speed Impeller, Backward Curve, 1260 CFM @ 0.000-In,Including HEPA filter , Including Carbon filter
Catalog No. 50-152-7918 Supplier Cleatech Supplier No. 11002FHC
Portable 32" Width Bench top ductless exhaust hood, Work space 31"Wx 17.75"Dx 17"H , Polypropylene worksurface, Static-dissipative PVC walls, Clear Static-dissipative PVC front tilt-up sash,Polypropylene Fan/ Filter Housing. 110VAC, 60Hz. 155W Adjustable speed Impeller, Backward Curve, 1260 CFM @ 0.000-In,Including HEPA filter , Including Carbon filter