Learn More
Cleatech GLOVE BOX W/ STND & AIRLOCK
SDP
Catalog No. 501527899
Encompass_Preferred
Two-Port Portable Mini Glovebox, non-vacuum, Clear Acrylic, 27"Wx 18"Dx 18"H, one swing up access door with opening of 8"x18"x8" Diameter glove ports with stainless steel clamp. Including 30" height, powder coated steel Stand ,Including Air Lock (transfer Chamber,11.5"Wx 10.5"Dx10.5"H), Including Gas In Port and Pressure relief Valve , Including 15 mil Chemical Resistance Gloves
Catalog No. 50-152-7899 Supplier Cleatech Supplier No. 22002AGGTS
Two-Port Portable Mini Glovebox, non-vacuum, Clear Acrylic, 27"Wx 18"Dx 18"H, one swing up access door with opening of 8"x18"x8" Diameter glove ports with stainless steel clamp. Including 30" height, powder coated steel Stand ,Including Air Lock (transfer Chamber,11.5"Wx 10.5"Dx10.5"H), Including Gas In Port and Pressure relief Valve , Including 15 mil Chemical Resistance Gloves