Learn More
Cleatech ESD SAFE ACRYLIC GLOVE BOX
SDP
Catalog No. 501527900
Encompass_Preferred
Two-Port Portable Mini Glovebox, non-vacuum, Static-Dissipative Acrylic Transparent, 27"Wx 18"Dx 18"H, one swing up access door with opening of 8"x18", 8" Diameter glove ports with stainless steel clamp, Including Gas In Port and Pressure relief Valve , Including 15 mil Chemical Resistance Gloves
Catalog No. 50-152-7900 Supplier Cleatech Supplier No. 22002EGG
Two-Port Portable Mini Glovebox, non-vacuum, Static-Dissipative Acrylic Transparent, 27"Wx 18"Dx 18"H, one swing up access door with opening of 8"x18", 8" Diameter glove ports with stainless steel clamp, Including Gas In Port and Pressure relief Valve , Including 15 mil Chemical Resistance Gloves