Learn More
Cleatech SD-AC GLOVE BOX W/ 30" STND
SDP
Catalog No. 501527901
Encompass_Preferred
Two-Port Portable Mini Glovebox, non-vacuum, Static-Dissipative Acrylic Transparent, 27"Wx 18"Dx 18"H, one swing up access door with opening of 8"x18", 8" Diameter glove ports with stainless steel clamp, Including 30" height, powder coated steel Stand , Including Gas In Port and Pressure relief Valve , Including 15 mil Chemical Resistance Gloves
Catalog No. 50-152-7901 Supplier Cleatech Supplier No. 22002EGGS
Two-Port Portable Mini Glovebox, non-vacuum, Static-Dissipative Acrylic Transparent, 27"Wx 18"Dx 18"H, one swing up access door with opening of 8"x18", 8" Diameter glove ports with stainless steel clamp, Including 30" height, powder coated steel Stand , Including Gas In Port and Pressure relief Valve , Including 15 mil Chemical Resistance Gloves