Learn More
Cleatech SD-AC GLOVE BOX W/ AIR LOCK
SDP
Catalog No. 501527902
Encompass_Preferred
Two-Port Portable Mini Glovebox, non-vacuum, Static-Dissipative Acrylic Transparent, 27"Wx 18"Dx 18"H, one swing up access door with opening of 8"x18", 8" Diameter glove ports with stainless steel clamp,Including Air Lock (transfer Chamber 11.5"Wx 10.5"Dx10.5"H) , Including Gas In Port and Pressure relief Valve , Including 15 mil Chemical Resistance Gloves
Catalog No. 50-152-7902 Supplier Cleatech Supplier No. 22002EGGT
Two-Port Portable Mini Glovebox, non-vacuum, Static-Dissipative Acrylic Transparent, 27"Wx 18"Dx 18"H, one swing up access door with opening of 8"x18", 8" Diameter glove ports with stainless steel clamp,Including Air Lock (transfer Chamber 11.5"Wx 10.5"Dx10.5"H) , Including Gas In Port and Pressure relief Valve , Including 15 mil Chemical Resistance Gloves