Learn More
Cleatech PP GLOVE BOX W/ 30" STAND
SDP
Catalog No. 501527905
Encompass_Preferred
Two-Port Portable Mini Glovebox, non-vacuum, White Polypropylene body, Clear Static Dissipative PVC front window, 27"Wx 18"Dx 18"H, one swing Up access door with opening of 8"x18", 8" Diameter glove ports with stainless steel clamp, Including built-in flourescent light 120V, Including 30" height, powder coated steel Stand , Including Gas In Port and Pressure relief Valve , Including 15 mil Chemical Resistance Gloves
Catalog No. 50-152-7905 Supplier Cleatech Supplier No. 22002DGGS
Two-Port Portable Mini Glovebox, non-vacuum, White Polypropylene body, Clear Static Dissipative PVC front window, 27"Wx 18"Dx 18"H, one swing Up access door with opening of 8"x18", 8" Diameter glove ports with stainless steel clamp, Including built-in flourescent light 120V, Including 30" height, powder coated steel Stand , Including Gas In Port and Pressure relief Valve , Including 15 mil Chemical Resistance Gloves