Learn More
Bachem Ac-Pro-Leu-Gly-OH
SDP
Supplier:  Bachem 4013355.1000
Encompass_Preferred
Ac-Pro-Leu-Gly-OH, 89626-38-0, C15H25N3O5, Mr 327.38, 1g. Cleavage product of MMP-1, -2 and -3 substrate Ac-Pro-Leu-Gly-psi[COS]Leu-Leu-Gly-OEt .
Catalog No. 50-194-6720