Learn More
Bachem H-Cit-AMC
SDP
Catalog No. 501947005
Encompass_Preferred
H-Cit-AMC Trifluoroacetate, 93753-78-7, C16H20N4O4, Mr 332.36, 1g. H-Orn(carbamoyl)-AMC. Cleaved by the cysteine proteases aleurain, a protease from barley related to cathepsin H, and bleomycin hydrolase.
Catalog No. 50-194-7005 Supplier Bachem Supplier No. 4006058.1000
Add to Cart
Add to Cart
Request a Quote
H-Cit-AMC Trifluoroacetate, 93753-78-7, C16H20N4O4, Mr 332.36, 1g. H-Orn(carbamoyl)-AMC. Cleaved by the cysteine proteases aleurain, a protease from barley related to cathepsin H, and bleomycin hydrolase.