Learn More
Abcam Recombinant human Cleaved Caspase-3 protein, 50UG
Supplier:  Abcam AB19844750UG
Encompass_Preferred
Recombinant human Cleaved Caspase-3 protein is a Human Full Length protein, in the 1 to 277 aa range, expressed in Escherichia coli, with >99% purity and suitable for SDS-PAGE, FuncS.
Protein/ Peptide/ Enzyme Synonyms: CPP32, CASP3, Caspase-3, CASP-3, Apopain, Cysteine protease CPP32, Protein Yama, SREBP cleavage activity 1, CPP-32, SCA-1
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2065036