Learn More
Cell Signaling Technology Cleaved Caspase-9 (Asp353) Polyclonal Antibody, Rabbit, Mouse and Rat, 100 μL
SDP
Catalog No. 502729854
Encompass
This antibody detects endogenous levels of the large fragment (17 kDa or 38 kDa with prodomain) of caspase-9 resulting from cleavage at aspartic acid 353. It does not recognize full-length caspase-9 or other cleaved caspases. The antibodies are generated by immunizing animals with a synthetic peptide corresponding to amino-terminal residues surrounding Asp353 of rat caspase-9 and are purified via protein A and peptide affinity chromatography.
  • Validated for western blotting and immunofluorescence applications.
  • Reacts with mouse and rat species.
  • Supplied in a formulation containing 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, and 50% glycerol.
  • Concentration is 17 μg/mL.
Catalog No. 50-272-9854 Supplier Cell Signaling Technology Supplier No. 9507S
This antibody detects endogenous levels of the large fragment (17 kDa or 38 kDa with prodomain) of caspase-9 resulting from cleavage at aspartic acid 353. It does not recognize full-length caspase-9 or other cleaved caspases. The antibodies are generated by immunizing animals with a synthetic peptide corresponding to amino-terminal residues surrounding Asp353 of rat caspase-9 and are purified via protein A and peptide affinity chromatography.
  • Validated for western blotting and immunofluorescence applications.
  • Reacts with mouse and rat species.
  • Supplied in a formulation containing 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, and 50% glycerol.
  • Concentration is 17 μg/mL.