Learn More
ABclonal Technology Cleaved Gasdermin DGly276
SDP
Supplier:  ABclonal Technology A26924100UL
Encompass_Preferred
Gasdermin D is a member of the gasdermin family Members of this family appear to play a role in regulation of epithelial proliferation Gasdermin D has been suggested to act as a tumor suppressor Alternatively spliced transcript variants have been described
Catalog No. 50-000-12205