Learn More
Abcam Cleaved Tau ELISA Kit (Human Asp738/Mouse Asp713), 1 x 96 Tests
Catalog No. 502656344
Encompass_Preferred
Cleaved Tau ELISA Kit (Human Asp738/Mouse Asp713) is a single-wash 90-min Simplestep used to quantify Cleaved Tau (Human Asp738/Mouse Asp713) with a sensitivity of 0.075 ng/mL. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-6344 Supplier Abcam Supplier No. AB2695571X96TESTS
Add to Cart
Add to Cart
Cleaved Tau ELISA Kit (Human Asp738/Mouse Asp713) is a single-wash 90-min Simplestep used to quantify Cleaved Tau (Human Asp738/Mouse Asp713) with a sensitivity of 0.075 ng/mL. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement