Learn More
Meso Scale Discovery Cleaved/Total CaspaseWCL20
SDP
Catalog No. 5000080603
Encompass
The Meso Scale Discovery Cleaved Total Caspase-3 Whole Cell Lysate Kit is designed to measure cleaved caspase-3 in cell lysates The kit contains a 96-well plate coated with a caspase-3 specific capture antibody a detection antibody conjugated to MSD SULFO-TAG and cell lysis buffer The assay uses electrochemiluminescence for high sensitivity detection of cleaved caspase-3 It can detect endogenous levels of caspase-3 and analyze caspase-3 activation and inhibition in response to apoptotic stimuli and caspase inhibitors The kit provides all necessary reagents for performing the assay in a 96-well format
Catalog No. 50-000-80603 Supplier Meso Scale Discovery Supplier No. K15140D3
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery Cleaved Total Caspase-3 Whole Cell Lysate Kit is designed to measure cleaved caspase-3 in cell lysates The kit contains a 96-well plate coated with a caspase-3 specific capture antibody a detection antibody conjugated to MSD SULFO-TAG and cell lysis buffer The assay uses electrochemiluminescence for high sensitivity detection of cleaved caspase-3 It can detect endogenous levels of caspase-3 and analyze caspase-3 activation and inhibition in response to apoptotic stimuli and caspase inhibitors The kit provides all necessary reagents for performing the assay in a 96-well format