Learn More
Meso Scale Discovery Cleaved/Total CaspaseWCLPl
SDP
Catalog No. 5000080553
Encompass
The Meso Scale Discovery Cleaved Total Caspase-3 Whole Cell Lysate Kit is designed to detect cleaved caspase-3 in cell lysates The kit contains a 96-well plate coated with a caspase-3 specific capture antibody a sulfo-tag labeled detection antibody buffers and calibrators The assay uses electrochemiluminescence detection to provide a sensitive and robust method for measuring caspase-3 activation during apoptosis This kit allows for high-throughput screening and monitoring of caspase-3 activation in cells during drug discovery and research applications The kit has a detection range of 50 - 50 000 pg/mL cleaved caspase-3
Catalog No. 50-000-80553 Supplier Meso Scale Discovery Supplier No. K15140D1
The Meso Scale Discovery Cleaved Total Caspase-3 Whole Cell Lysate Kit is designed to detect cleaved caspase-3 in cell lysates The kit contains a 96-well plate coated with a caspase-3 specific capture antibody a sulfo-tag labeled detection antibody buffers and calibrators The assay uses electrochemiluminescence detection to provide a sensitive and robust method for measuring caspase-3 activation during apoptosis This kit allows for high-throughput screening and monitoring of caspase-3 activation in cells during drug discovery and research applications The kit has a detection range of 50 - 50 000 pg/mL cleaved caspase-3