Learn More
Electron Microscopy Sciences Small Sample Wafer Cleaving Pliers
SDP
Catalog No. 501928732
Encompass_Preferred
Custom, handheld cleaving pliers cleave small samples into pieces from 1 - 30 mm. Stainless steel covered with plastic handles for strong grip. Soft nylon covered jaws that do not damage your sample. Good for thin and small samples. Lightweight and easy to use.
Catalog No. 50-192-8732 Supplier Electron Microscopy Sciences Supplier No. 7647
Custom, handheld cleaving pliers cleave small samples into pieces from 1 - 30 mm. Stainless steel covered with plastic handles for strong grip. Soft nylon covered jaws that do not damage your sample. Good for thin and small samples. Lightweight and easy to use.