Learn More
Molex Molex-Polymicro Cleaving Stone W/Molex Logo
Supplier:  Molex 1068680064/DUP
Encompass
This item has a minimum qty of 20 per supplier requirements.
Cleaving Stone, 1" x 1" x 1/32 Overall Dimensions, One Piece Per Bag; Ideal for Hand Cleaving Capillary Tubing
Catalog No. NC1279124