Learn More
ACROBiosystems ACROBiosystems Human CLEC12A / MICL / CLL-1 Protein, Fc Tag
Catalog No. 501956527
Encompass_Preferred
Recombinant Protein;50UG;Human CLEC12A, Fc Tag (CLA-H5266) is expressed from human 293 cells (HEK293). It contains AA His 65 - Ala 265 (Accession # Q5QGZ9-2).;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 50.2 kDa. The protein migrates as 55-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6527 Supplier ACROBiosystems Supplier No. CLAH526650UG
Recombinant Protein;50UG;Human CLEC12A, Fc Tag (CLA-H5266) is expressed from human 293 cells (HEK293). It contains AA His 65 - Ala 265 (Accession # Q5QGZ9-2).;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 50.2 kDa. The protein migrates as 55-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.