Learn More
ACROBiosystems ACROBiosystems Human CLEC12A / MICL / CLL-1 Protein, His Tag
Catalog No. 501956480
Encompass_Preferred
Recombinant Protein;50UG;Human CLEC12A, His Tag (CLA-H5245) is expressed from human 293 cells (HEK293). It contains AA His 75 - Ala 275 (Accession # NP_001193939.1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 25.6 kDa. The protein migrates as 35-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6480 Supplier ACROBiosystems Supplier No. CLAH524550UG
Recombinant Protein;50UG;Human CLEC12A, His Tag (CLA-H5245) is expressed from human 293 cells (HEK293). It contains AA His 75 - Ala 275 (Accession # NP_001193939.1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 25.6 kDa. The protein migrates as 35-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.