Learn More
Genscript Corporation CLEC12A/MICL/CLL-1 hFc Chimera
Supplier:  Genscript Corporation Z03960100
Encompass
CLEC12A has recently been identified as an antigen expressed on leukemic stem cells and leukemic blasts Given the fact that this expression profile seems stable throughout diagnosis treatment and relapse on leukemic blasts and leukemic stem cells CLEC12A can be considered a highly potent and reliable marker for the detection of measurable residual disease and therefore applicable for risk stratification and prognostication in AML
Catalog No. 50-000-56759