Learn More
Medchemexpress LLC CLEC12A/MICL Cynomo 50ug | 50ug
SDP
Catalog No. 5000187775
Encompass_Preferred
CLEC12A/MICL Protein is a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily CLEC12A is a cell surface receptor known to regulate signaling cascades and facilitate the tyrosine phosphorylation of target MAP kinases It interacts with PTPN6 and PTPN11 indicating its involvement in intricate cellular signaling processes CLEC12A/MICL Protein Cynomolgus (HEK293 His) is the recombinant cynomolgus-derived CLEC12A/MICL protein expressed by HEK293 with N-His labeled tag
Catalog No. 50-001-87775 Supplier Medchemexpress LLC Supplier No. HYP7790350UG
CLEC12A/MICL Protein is a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily CLEC12A is a cell surface receptor known to regulate signaling cascades and facilitate the tyrosine phosphorylation of target MAP kinases It interacts with PTPN6 and PTPN11 indicating its involvement in intricate cellular signaling processes CLEC12A/MICL Protein Cynomolgus (HEK293 His) is the recombinant cynomolgus-derived CLEC12A/MICL protein expressed by HEK293 with N-His labeled tag