Learn More
Aviva Systems Biology Corp Clec1b Antibody - N-terminal region (ARP47260_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47260_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Clec1b. It was validated on Western Blot. . Alternate names: 1810061I13Rik, AI465458, Clec-2, Clec2, mCLEC-2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-285-293