Learn More
Genscript Corporation CLEC2D hFc ChimeraBiotin Av
Catalog No. 5000059095
Encompass
C-type lectin domain family 2 member D (CLEC2D) is implicated in the immune response Pre-eclampsia and HIV infection have opposing immune responses The contrasting expression of CLEC2D in HIV infection and pre-eclampsia is demonstrative of the immunosuppressive and pro-inflammatory roles of the respective pathologies
Catalog No. 50-000-59095 Supplier Genscript Corporation Supplier No. Z04434100
C-type lectin domain family 2 member D (CLEC2D) is implicated in the immune response Pre-eclampsia and HIV infection have opposing immune responses The contrasting expression of CLEC2D in HIV infection and pre-eclampsia is demonstrative of the immunosuppressive and pro-inflammatory roles of the respective pathologies