Learn More
Sino Biological Recombinant Human Prolactin Receptor Protein (His Tag)
Supplier:  Sino Biological 11476MM02100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human PRLR (NP_000940.1) (Met 1-Asp 234) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-8447