Learn More
Sino Biological CLEC4D Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 11485MM04100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CLEC4D (rh CLEC4D; Catalog#11485-H07H; NP_525126.2; Gly52-Asn215). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-8631