Learn More
Aviva Systems Biology Corp CLEC4M Antibody - N-terminal region (ARP42396_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42396_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CLEC4M. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human CLEC4M. Alternate names: CD209L, CD299, DC-SIGN2, DC-SIGNR, DCSIGNR, HP10347, L-SIGN, LSIGN, MGC129964, MGC47866ApplicationIHC, WB. Species reactivity: Human, Pig
Catalog No. 50-014-58212