Learn More
Aviva Systems Biology Corp CLEC4M Antibody - middle region (ARP42752_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42752_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CLEC4M. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human CLEC4M. Alternate names: CD299, LSIGN, CD209L, L-SIGN, DCSIGNR, HP10347, DC-SIGN2, DC-SIGNR, MGC47866, MGC129964, CLEC4MApplicationWB. Species reactivity: Horse, Human, Pig
Catalog No. 50-285-021