Learn More
Aviva Systems Biology Corp CLEC5A Antibody - C-terminal region (ARP62114_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62114_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CLEC5A. It was validated on Western Blot. . Alternate names: CLECSF5, MDL-1, MDL1, MGC138304ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rat, Yeast
Catalog No. 50-287-575