Learn More
NEOGEN Cystine Lactose Electrolyte Deficient (CLED) Agar, 500 g
SDP
Catalog No. 502015252
Encompass_Preferred
Cystine Lactose Electrolyte Deficient (CLED) Agar is used for the differentiation and enumeration of microorganisms in urine. A medium for urine culture where the absence of electrolytes inhibits the swarming of Proteus spp. Cystine is added for the benefit of those organisms which have a specific cystine requirement. Differentiation of lactose and non-lactose fermenters is achieved using bromothymol blue as pH indicator. This medium supports the growth of Streptococcus pyogenes and most other fastidious organisms that do not require blood.  Formula:Balanced Peptone - 4.0 g/LBeef Extract - 3.0 g/LTryptone - 4.0 g/LLactose - 10.0 g/LL-Cystine - 0.128 g/LBromothymol Blue - 0.02 g/LAgar - 15.0 g/LFor more information visit https://www.neogen.com/solutions/Microbiology/cystine-lactose-electrolyte-deficient-agar
Catalog No. 50-201-5252 Supplier NEOGEN Supplier No. 700003061
Cystine Lactose Electrolyte Deficient (CLED) Agar is used for the differentiation and enumeration of microorganisms in urine. A medium for urine culture where the absence of electrolytes inhibits the swarming of Proteus spp. Cystine is added for the benefit of those organisms which have a specific cystine requirement. Differentiation of lactose and non-lactose fermenters is achieved using bromothymol blue as pH indicator. This medium supports the growth of Streptococcus pyogenes and most other fastidious organisms that do not require blood.  Formula:Balanced Peptone - 4.0 g/LBeef Extract - 3.0 g/LTryptone - 4.0 g/LLactose - 10.0 g/LL-Cystine - 0.128 g/LBromothymol Blue - 0.02 g/LAgar - 15.0 g/LFor more information visit https://www.neogen.com/solutions/Microbiology/cystine-lactose-electrolyte-deficient-agar