Learn More
Medchemexpress LLC Cleistanthin B | 30021-77-3 | 98.2% | 542.49 | 1 MG
SDP
Catalog No. 5000426869
Encompass_Preferred
Cleistanthin B is an orally active arylnaphthalene lignan lactone glycoside that demonstrates anti-SARS-CoV-2 effects in Vero cells and exhibits antitumor, diuretic, and antihypertensive properties in vivo.
  • Exhibits anti-SARS-CoV-2 effects in Vero cells with an EC50 of 6.51 μM
  • Shows no cytotoxicity to Vero cells in concentrations up to 100 μM
  • Demonstrates antitumor activity against EAC and DAL tumor mice
  • Also shows diuretic and antihypertensive effects
Catalog No. 50-004-26869 Supplier Medchemexpress LLC Supplier No. HYN93511MG
Cleistanthin B is an orally active arylnaphthalene lignan lactone glycoside that demonstrates anti-SARS-CoV-2 effects in Vero cells and exhibits antitumor, diuretic, and antihypertensive properties in vivo.
  • Exhibits anti-SARS-CoV-2 effects in Vero cells with an EC50 of 6.51 μM
  • Shows no cytotoxicity to Vero cells in concentrations up to 100 μM
  • Demonstrates antitumor activity against EAC and DAL tumor mice
  • Also shows diuretic and antihypertensive effects