Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Clemastine-d5 fumarate | 14976-57-9 | 99.5% | 464.99 | C25H25D5ClNO5 | 10 MG
SDP

Catalog No. 5000297402
Encompass_Preferred

Clemastine-d5 fumarate is the deuterium-labeled (d5) analogue of clemastine fumarate supplied as a stable-isotope analytical standard for research and quantitative mass spectrometry. It is intended for use as an internal standard or tracer in LC-MS/MS and other analytical workflows, and is not for clinical use.

  • Deuterium-labeled (d5) stable isotope standard.
  • Intended for research and analytical use only.
  • High purity (≈99.5%).
  • Molecular weight 464.99 g/mol.
  • Chemical formula C25H25D5ClNO5.
  • Available in multiple pack sizes, including 10 mg.

Catalog No. 50-002-97402 Supplier Medchemexpress LLC Supplier No. HYB0298AS10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Clemastine-d5 fumarate is the deuterium-labeled (d5) analogue of clemastine fumarate supplied as a stable-isotope analytical standard for research and quantitative mass spectrometry. It is intended for use as an internal standard or tracer in LC-MS/MS and other analytical workflows, and is not for clinical use.

  • Deuterium-labeled (d5) stable isotope standard.
  • Intended for research and analytical use only.
  • High purity (≈99.5%).
  • Molecular weight 464.99 g/mol.
  • Chemical formula C25H25D5ClNO5.
  • Available in multiple pack sizes, including 10 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.