Learn More
Apexbio Technology LLC Clemastine Fumarate 14976-57-9 100mg
SDP
Catalog No. 5000089928
Encompass_Preferred
Clemastine Fumarate (CAS 14976-57-9) is a small molecule that functions as a selective antagonist of the histamine H1 receptor It demonstrates high affinity for the receptor with an inhibitory concentration (IC50) of 3 nM By blocking histamine H1 receptor-mediated signaling clemastine fumarate is utilized in research to study histaminergic pathways and allergic responses It is also employed to investigate the pharmacological modulation of H1 receptor activity in various cellular and in vivo models
Catalog No. 50-000-89928 Supplier Apexbio Technology LLC Supplier No. B1558100
Clemastine Fumarate (CAS 14976-57-9) is a small molecule that functions as a selective antagonist of the histamine H1 receptor It demonstrates high affinity for the receptor with an inhibitory concentration (IC50) of 3 nM By blocking histamine H1 receptor-mediated signaling clemastine fumarate is utilized in research to study histaminergic pathways and allergic responses It is also employed to investigate the pharmacological modulation of H1 receptor activity in various cellular and in vivo models