Learn More
Medchemexpress LLC Clemastine fumarate | 14976-57-9 | 99.96% | 1 ML
SDP
Catalog No. 5000405876
Encompass_Preferred
Clemastine fumarate is an orally active, blood-brain barrier-permeable H1 histamine receptor antagonist with potent antiallergic effects. It also antagonizes muscarinic acetylcholine receptors (mAChR), particularly the M1 and M4 subtypes. Beyond antihistamine effects, Clemastine fumarate shows various pharmacological activities.
  • Promoting remyelination in the central nervous system
  • Activating autophagy and pyroptosis
  • Exerting anti-apoptotic and neuroprotective effects
  • Suppressing inflammation
Catalog No. 50-004-05876 Supplier Medchemexpress LLC Supplier No. HYB0298A10MM/1ML
Clemastine fumarate is an orally active, blood-brain barrier-permeable H1 histamine receptor antagonist with potent antiallergic effects. It also antagonizes muscarinic acetylcholine receptors (mAChR), particularly the M1 and M4 subtypes. Beyond antihistamine effects, Clemastine fumarate shows various pharmacological activities.
  • Promoting remyelination in the central nervous system
  • Activating autophagy and pyroptosis
  • Exerting anti-apoptotic and neuroprotective effects
  • Suppressing inflammation