Learn More
Selleck Chemical LLC Clemastine fumarate S1847-1g
SDP
Catalog No. 503109592
Encompass_Preferred
Clemastine fumarate is a selective histamine H1 receptor antagonist with IC50 of 3 nM Clemastine stimulates autophagy via mTOR pathway
Catalog No. 50-310-9592 Supplier Selleck Chemical LLC Supplier No. S18471G
Clemastine fumarate is a selective histamine H1 receptor antagonist with IC50 of 3 nM Clemastine stimulates autophagy via mTOR pathway