Learn More
Cayman Chemical Clemastine (fumarate), Purity: ≥98%, CAS: 14976-57-9, 100 mg
SDP
Catalog No. 502973076
Encompass_Preferred
A histamine H1 receptor antagonist (Ki = 0.26 nM); selective for the histamine H1 receptor over mAChRs (Ki = 16 nM), as well as over σ1 and σ2 and α2A-, α2B-, and α2C-adrenergic receptors (Kis = 67, 15, 1,365, 691, and 545 nM, respectively); reduces ovalbumin-induced bronchial obstruction in ovalbumin-sensitized guinea pigs alone or in combination with cimetidine at 0.12 mg/kg; decreases SARS-CoV-2 replication in SARS-CoV-2-infected Vero E6 cells (EC50 = 0.95 µM)
Catalog No. 50-297-3076 Supplier Cayman Chemical Supplier No. 14637100 MG
A histamine H1 receptor antagonist (Ki = 0.26 nM); selective for the histamine H1 receptor over mAChRs (Ki = 16 nM), as well as over σ1 and σ2 and α2A-, α2B-, and α2C-adrenergic receptors (Kis = 67, 15, 1,365, 691, and 545 nM, respectively); reduces ovalbumin-induced bronchial obstruction in ovalbumin-sensitized guinea pigs alone or in combination with cimetidine at 0.12 mg/kg; decreases SARS-CoV-2 replication in SARS-CoV-2-infected Vero E6 cells (EC50 = 0.95 µM)