Learn More
Apexbio Technology LLC Clemizole hydrochloride 1163-36-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000064132
Encompass_Preferred
Clemizole hydrochloride is a histamine H1 receptor antagonist reported to inhibit hepatitis C virus (HCV) replication by targeting the viral NS4B protein a component essential for viral RNA replication Clemizole demonstrates antiviral activity in vitro against HCV genotype 2a subgenomic replicons exhibiting an IC50 value around 8 mM When combined with HCV protease inhibitors (such as VX950 or SCH503034) clemizole displays synergistic antiviral effects In contrast additive antiviral effects occur in combination with interferon ribavirin or polymerase inhibitors Clemizole is utilized in research contexts aimed at evaluating its efficacy potential antiviral combinations metabolism pathways and pharmacokinetics relevant to its consideration as a therapeutic candidate for HCV infection
Catalog No. 50-000-64132 Supplier Apexbio Technology LLC Supplier No. A33165.1
Clemizole hydrochloride is a histamine H1 receptor antagonist reported to inhibit hepatitis C virus (HCV) replication by targeting the viral NS4B protein a component essential for viral RNA replication Clemizole demonstrates antiviral activity in vitro against HCV genotype 2a subgenomic replicons exhibiting an IC50 value around 8 mM When combined with HCV protease inhibitors (such as VX950 or SCH503034) clemizole displays synergistic antiviral effects In contrast additive antiviral effects occur in combination with interferon ribavirin or polymerase inhibitors Clemizole is utilized in research contexts aimed at evaluating its efficacy potential antiviral combinations metabolism pathways and pharmacokinetics relevant to its consideration as a therapeutic candidate for HCV infection