Learn More
Medchemexpress LLC Clemizole-d4 | 442-52-4 | 99.8% | 329.86 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY30234S1MG
Encompass_Preferred
Clemizole-d4 is a deuterium labeled Clemizole. Clemizole is an H1 histamine receptor antagonist that has been found to substantially inhibit HCV replication.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-04326