Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Clemizole-d4 | 442-52-4 | 99.8% | 329.86 | C19H16D4ClN3 | 10 MG
SDP

Catalog No. 5000297589
Encompass_Preferred

Clemizole-d4 is a deuterium-labeled analogue of clemizole supplied as a high-purity solid for use as an internal standard and tracer in quantitative analytical methods. It is suitable for studies involving histamine H1 receptor activity and for research into HCV replication pathways, and is prepared to support reproducible mass spectrometry and NMR measurements.

  • Used as an internal standard and tracer for NMR, GC-MS, and LC-MS analyses.
  • High purity (99.82%) suitable for quantitative work.
  • Molecular weight 329.86; formula C19H16D4ClN3.
  • White to off-white solid with recommended frozen storage for long-term stability.
  • Soluble in DMSO at 100 mg/mL; may require warming and sonication.
  • Intended for research use only; not for human or diagnostic use.

Catalog No. 50-002-97589 Supplier Medchemexpress LLC Supplier No. HY30234S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Clemizole-d4 is a deuterium-labeled analogue of clemizole supplied as a high-purity solid for use as an internal standard and tracer in quantitative analytical methods. It is suitable for studies involving histamine H1 receptor activity and for research into HCV replication pathways, and is prepared to support reproducible mass spectrometry and NMR measurements.

  • Used as an internal standard and tracer for NMR, GC-MS, and LC-MS analyses.
  • High purity (99.82%) suitable for quantitative work.
  • Molecular weight 329.86; formula C19H16D4ClN3.
  • White to off-white solid with recommended frozen storage for long-term stability.
  • Soluble in DMSO at 100 mg/mL; may require warming and sonication.
  • Intended for research use only; not for human or diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.