Learn More
Medchemexpress LLC Clemizole hydrochloride | 1163-36-6 | 100.0% | 362.30 | 50 MG
SDP
Catalog No. 5000405237
Encompass_Preferred
Clemizole hydrochloride is an H1 histamine receptor antagonist that has been shown to substantially inhibit HCV replication. It also acts as an inhibitor of the TRPC5 channel. The IC50 of Clemizole hydrochloride for RNA binding by NS4B is 24 nM, while its EC50 for viral replication is 8 μM.
  • Inhibits HCV RNA replication in cell culture.
  • Suppresses NS4B's RNA binding.
  • Offers minimal host cell toxicity.
  • Functions as a novel inhibitor of TRPC5 channels.
  • Efficiently blocks TRPC5 currents and Ca2+ entry in the low micromolar range.
  • Exhibits six-fold selectivity for TRPC5 over TRPC4β.
  • Displays almost 10-fold selectivity over TRPC3 and TRPC6.
Catalog No. 50-004-05237 Supplier Medchemexpress LLC Supplier No. HY30234A50MG
Clemizole hydrochloride is an H1 histamine receptor antagonist that has been shown to substantially inhibit HCV replication. It also acts as an inhibitor of the TRPC5 channel. The IC50 of Clemizole hydrochloride for RNA binding by NS4B is 24 nM, while its EC50 for viral replication is 8 μM.
  • Inhibits HCV RNA replication in cell culture.
  • Suppresses NS4B's RNA binding.
  • Offers minimal host cell toxicity.
  • Functions as a novel inhibitor of TRPC5 channels.
  • Efficiently blocks TRPC5 currents and Ca2+ entry in the low micromolar range.
  • Exhibits six-fold selectivity for TRPC5 over TRPC4β.
  • Displays almost 10-fold selectivity over TRPC3 and TRPC6.