Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Clemizole hydrochloride | 1163-36-6 | 99.99% | 362.30 | 100 MG
SDP

Catalog No. 5000404817
Encompass_Preferred

Clemizole hydrochloride is an H1 histamine receptor antagonist that substantially inhibits HCV replication and is an inhibitor of the TRPC5 channel. It has an IC50 of 24 nM for RNA binding by NS4B and an EC50 of 8 μM for viral replication. For research use only.

  • H1 histamine receptor antagonist.
  • Substantially inhibits HCV replication.
  • Inhibitor of TRPC5 channel.
  • EC50 of 8 μM for viral replication.
  • IC50 of 24 nM for RNA binding by NS4B.
  • Inhibits HCV RNA replication in cell culture mediated by suppression of NS4B's RNA binding with little toxicity for the host cell.
  • Efficiently blocks TRPC5 currents and Ca2+ entry in the low micromolar range (IC50=1.0-1.3 μM).
  • Exhibits six-fold selectivity for TRPC5 over TRPC4β (IC50=6.4 μM) and almost 10-fold selectivity over TRPC3 (IC50=9.1 μM) and TRPC6 (IC50=11.3 μM).

Catalog No. 50-004-04817 Supplier Medchemexpress LLC Supplier No. HY30234A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Clemizole hydrochloride is an H1 histamine receptor antagonist that substantially inhibits HCV replication and is an inhibitor of the TRPC5 channel. It has an IC50 of 24 nM for RNA binding by NS4B and an EC50 of 8 μM for viral replication. For research use only.

  • H1 histamine receptor antagonist.
  • Substantially inhibits HCV replication.
  • Inhibitor of TRPC5 channel.
  • EC50 of 8 μM for viral replication.
  • IC50 of 24 nM for RNA binding by NS4B.
  • Inhibits HCV RNA replication in cell culture mediated by suppression of NS4B's RNA binding with little toxicity for the host cell.
  • Efficiently blocks TRPC5 currents and Ca2+ entry in the low micromolar range (IC50=1.0-1.3 μM).
  • Exhibits six-fold selectivity for TRPC5 over TRPC4β (IC50=6.4 μM) and almost 10-fold selectivity over TRPC3 (IC50=9.1 μM) and TRPC6 (IC50=11.3 μM).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.