Learn More
Medchemexpress LLC Clemizole hydrochloride | 1163-36-6 | MFCD00051435 | 100.0% | 362.30 | C19H21Cl2N3 | 10 MG
SDP
Catalog No. 5000297739
Encompass_Preferred
Clemizole hydrochloride is a research-grade H1 histamine receptor antagonist with reported antiviral activity against hepatitis C virus and inhibitory effects on TRPC5 channels. It is supplied as a solid salt and as a solution formulation for biochemical and cell-based assays, and is commonly used in pharmacology and target-validation studies.
  • H1 histamine receptor antagonist activity
  • Inhibits hepatitis C virus replication in cell-based assays
  • Blocks TRPC5 ion channel activity
  • High purity reported at 99.99%
  • Supplied in small-mass vials and as a DMSO solution for assay use
  • Suitable for biochemical, pharmacology, and cell-based studies
Catalog No. 50-002-97739 Supplier Medchemexpress LLC Supplier No. HY30234A10MG
Clemizole hydrochloride is a research-grade H1 histamine receptor antagonist with reported antiviral activity against hepatitis C virus and inhibitory effects on TRPC5 channels. It is supplied as a solid salt and as a solution formulation for biochemical and cell-based assays, and is commonly used in pharmacology and target-validation studies.
  • H1 histamine receptor antagonist activity
  • Inhibits hepatitis C virus replication in cell-based assays
  • Blocks TRPC5 ion channel activity
  • High purity reported at 99.99%
  • Supplied in small-mass vials and as a DMSO solution for assay use
  • Suitable for biochemical, pharmacology, and cell-based studies