Learn More
Medchemexpress LLC HY-30234A 5mg Medchemexpress, Clemizole (hydrochloride) CAS:1163-36-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY30234A5MG
Encompass_Preferred
Medchemexpress, HY-30234A 5mg Clemizole (hydrochloride) CAS:1163-36-6 Clemizole hydrochloride is an H1 histamine receptor antagonist, is found to substantially inhibit HCV replication. The IC50 of Clemizole for RNA binding by NS4B is 24±1 nM, whereas its EC50 for viral replication is 8 µM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2225